Benzyl (R)-(-)-glycidyl ether 98%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD0151-25GEA
210
DKK
BLDPBD0151-25G
BLDPBD0151-1G
BLDPBD0151-10G
BLDPBD0151-5G
Benzyl (R)-(-)-glycidyl ether 98%
Benzyl (R)-(-)-glycidyl ether
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₁₀H₁₂O₂ Molekylvægt: 164,20 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00068409 CAS-nummer: 14618-80-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...