N1-(3,5-Di-tert-butylphenyl)benzene-1,2-diamine 95%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD01499775-250EA
504
DKK
BLDPBD01499775-250
N1-(3,5-Di-tert-butylphenyl)benzene-1,2-diamine 95%
N1-(3,5-Di-tert-butylphenyl)benzene-1,2-diamine
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formel:
C₂₀H₂₈N₂ Molekylvægt: 296,44 g/mol Opbevaringstemperatur: Stue temperatur |
CAS-nummer:
2365501-07-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...