tert-Butyl 3-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydropyridin-1(2H)-yl)propanoate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01491953-250EA 5840 DKK
BLDPBD01491953-250 BLDPBD01491953-1G BLDPBD01491953-100
tert-Butyl 3-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydropyridin-1(2H)-yl)propanoate 95%
tert-Butyl 3-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydropyridin-1(2H)-yl)propanoate

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₁₈H₃₂BNO₄
Molekylvægt: 337,26 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD32659969
CAS-nummer: 1802892-92-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR