3,5-Dichloro-4-(hydroxymethyl)benzamide 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01485088-250EA 3760 DKK
BLDPBD01485088-250 BLDPBD01485088-1G BLDPBD01485088-50M BLDPBD01485088-100
3,5-Dichloro-4-(hydroxymethyl)benzamide 95%
3,5-Dichloro-4-(hydroxymethyl)benzamide

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₇Cl₂NO₂
Molekylvægt: 220,0527 g/mol
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 2384505-95-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR