N-(4-(Naphthalen-1-yl)phenyl)-[1,1':2',1''-terphenyl]-4'-amine 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01481350-1GEA 590 DKK
BLDPBD01481350-1G BLDPBD01481350-250 BLDPBD01481350-5G
N-(4-(Naphthalen-1-yl)phenyl)-[1,1':2',1''-terphenyl]-4'-amine 98%
N-(4-(Naphthalen-1-yl)phenyl)-[1,1':2',1''-terphenyl]-4'-amine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₃₄H₂₅N
Molekylvægt: 447,56 g/mol
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 2762417-19-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR