3-((4-Bromo-1H-pyrazol-1-yl)methyl)isoxazole 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01480070-250EA 2300 DKK
BLDPBD01480070-250 BLDPBD01480070-100 BLDPBD01480070-1G
3-((4-Bromo-1H-pyrazol-1-yl)methyl)isoxazole 95%
3-((4-Bromo-1H-pyrazol-1-yl)methyl)isoxazole

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₆BrN₃O
Molekylvægt: 228,04 g/mol
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 1871725-17-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR