2-(4-((4-Chlorobenzyl)oxy)-3,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01479349-100EA 303 DKK
BLDPBD01479349-100
2-(4-((4-Chlorobenzyl)oxy)-3,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
2-(4-((4-Chlorobenzyl)oxy)-3,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Storage condition: Sealed in dry, Room Temperature

Formel: C₂₁H₂₆BClO₃
Molekylvægt: 372,69 g/mol
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 2828440-16-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR