O-(5,6,7,8-Tetrahydronaphthalen-2-yl) carbonochloridothioate 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01474385-25GEA 442 DKK
BLDPBD01474385-25G BLDPBD01474385-5G
O-(5,6,7,8-Tetrahydronaphthalen-2-yl) carbonochloridothioate 98%
O-(5,6,7,8-Tetrahydronaphthalen-2-yl) carbonochloridothioate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₁H₁₁ClOS
Molekylvægt: 226,72 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD20487682
CAS-nummer: 84995-63-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR