Dibromo(4,4'-dimethoxy-2,2'-bipyridine-κN1,κN1')Nickel 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01467563-1GEA 931 DKK
BLDPBD01467563-1G BLDPBD01467563-250 BLDPBD01467563-5G
Dibromo(4,4'-dimethoxy-2,2'-bipyridine-κN1,κN1')Nickel 95%
Dibromo(4,4'-dimethoxy-2,2'-bipyridine-κN1,κN1')Nickel

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₂H₁₂Br₂N₂NiO₂
Molekylvægt: 434,7373 g/mol
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 2409691-04-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR