tert-Butyl ((3-azabicyclo[3.1.0]hexan-6-yl)methyl)(methyl)carbamate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01463444-50MEA 2500 DKK
BLDPBD01463444-50M BLDPBD01463444-100 BLDPBD01463444-1G BLDPBD01463444-250
tert-Butyl ((3-azabicyclo[3.1.0]hexan-6-yl)methyl)(methyl)carbamate 95%
tert-Butyl ((3-azabicyclo[3.1.0]hexan-6-yl)methyl)(methyl)carbamate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₁₂H₂₂N₂O₂
Molekylvægt: 226,31 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD28634726
CAS-nummer: 1781667-67-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR