(4-Chloro-3-(methoxycarbonyl)-5-methylphenyl)boronic acid 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01460724-100EA 303 DKK
BLDPBD01460724-100 BLDPBD01460724-1G BLDPBD01460724-5G BLDPBD01460724-250
(4-Chloro-3-(methoxycarbonyl)-5-methylphenyl)boronic acid 97%
(4-Chloro-3-(methoxycarbonyl)-5-methylphenyl)boronic acid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₉H₁₀BClO₄
Molekylvægt: 228,4373 g/mol
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 2828440-15-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR