(1-(tert-Butoxycarbonyl)-1H-pyrrolo[2,3-b]pyridin-4-yl)boronic acid 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01459557-250EA 512 DKK
BLDPBD01459557-250 BLDPBD01459557-100
(1-(tert-Butoxycarbonyl)-1H-pyrrolo[2,3-b]pyridin-4-yl)boronic acid 95%
(1-(tert-Butoxycarbonyl)-1H-pyrrolo[2,3-b]pyridin-4-yl)boronic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₂H₁₅BN₂O₄
Molekylvægt: 262,0695 g/mol
Opbevaringstemperatur: Køleskab
CAS-nummer: 2159138-80-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR