(4-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)(morpholino)methanone 97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD01459464-100EA
303
DKK
BLDPBD01459464-100
BLDPBD01459464-250
BLDPBD01459464-1G
(4-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)(morpholino)methanone 97%
(4-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)(morpholino)methanone
Storage condition: Sealed in dry, 2-8°C
Formel:
C₁₇H₂₃BFNO₄ Molekylvægt: 335,17 g/mol Opbevaringstemperatur: Køleskab |
CAS-nummer:
2828440-06-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...