Methyl (S)-2-amino-3-(3,4-difluorophenyl)propanoate hydrochloride 95% (by HPLC)

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01456733-1GEA 5590 DKK
BLDPBD01456733-1G BLDPBD01456733-250 BLDPBD01456733-100
Methyl (S)-2-amino-3-(3,4-difluorophenyl)propanoate hydrochloride 95% (by HPLC)
Methyl (S)-2-amino-3-(3,4-difluorophenyl)propanoate hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₀H₁₂ClF₂NO₂
Molekylvægt: 251,6576 g/mol
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 2230086-42-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR