(S)-3-(3-Bromo-5-chlorophenyl)morpholine 95% (by HPLC)

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01455166-250EA 6730 DKK
BLDPBD01455166-250 BLDPBD01455166-100 BLDPBD01455166-50M
(S)-3-(3-Bromo-5-chlorophenyl)morpholine 95% (by HPLC)
(S)-3-(3-Bromo-5-chlorophenyl)morpholine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₀H₁₁BrClNO
Molekylvægt: 276,55 g/mol
Opbevaringstemperatur: Køleskab
CAS-nummer: 1344632-02-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR