5,5-Dimethyl-3-nitro-6,7-dihydro-5H-pyrazolo[5,1-b][1,3]oxazine 95+%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01454136-100EA 442 DKK
BLDPBD01454136-100 BLDPBD01454136-250
5,5-Dimethyl-3-nitro-6,7-dihydro-5H-pyrazolo[5,1-b][1,3]oxazine 95+%
5,5-Dimethyl-3-nitro-6,7-dihydro-5H-pyrazolo[5,1-b][1,3]oxazine

Storage condition: Inert atmosphere, 2-8°C

Formel: C₈H₁₁N₃O₃
Molekylvægt: 197,19 g/mol
Opbevaringstemperatur: Køleskab
CAS-nummer: 2170751-30-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR