Methyl octahydro-1H-quinolizine-9a-carboxylate 98% + (by GC)

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01452878-1GEA 2070 DKK
BLDPBD01452878-1G BLDPBD01452878-100 BLDPBD01452878-250
Methyl octahydro-1H-quinolizine-9a-carboxylate 98% + (by GC)
Methyl octahydro-1H-quinolizine-9a-carboxylate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₁H₁₉NO₂
Molekylvægt: 197,27 g/mol
Opbevaringstemperatur: Køleskab
CAS-nummer: 30832-17-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR