2-(4-Bromophenyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione 98%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01449198-5GEA 690 DKK
BLDPBD01449198-5G BLDPBD01449198-25G BLDPBD01449198-1G BLDPBD01449198-250
2-(4-Bromophenyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione 98%
2-(4-Bromophenyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₁H₁₁BBrNO₄
Molekylvægt: 311,924 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11215230
CAS-nummer: 943552-04-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR