(E)-Tetradec-5-en-1-ol 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01449190-100EA 1250 DKK
BLDPBD01449190-100 BLDPBD01449190-250 BLDPBD01449190-1G
(E)-Tetradec-5-en-1-ol 95%
(E)-Tetradec-5-en-1-ol

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₄H₂₈O
Molekylvægt: 212,37 g/mol
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 62936-14-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR