7-Fluoro-5-methoxyquinoline-3-carboxylic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01448989-250EA 2900 DKK
BLDPBD01448989-250 BLDPBD01448989-1G BLDPBD01448989-100
7-Fluoro-5-methoxyquinoline-3-carboxylic acid 98%
7-Fluoro-5-methoxyquinoline-3-carboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₁H₈FNO₃
Molekylvægt: 221,1845 g/mol
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 2828438-97-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR