tert-Butyl (5,6,7,8-tetrahydroimidazo[1,2-a]pyridin-7-yl)carbamate 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01446189-250EA 2900 DKK
BLDPBD01446189-250 BLDPBD01446189-1G BLDPBD01446189-100
tert-Butyl (5,6,7,8-tetrahydroimidazo[1,2-a]pyridin-7-yl)carbamate 98%
tert-Butyl (5,6,7,8-tetrahydroimidazo[1,2-a]pyridin-7-yl)carbamate

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₂H₁₉N₃O₂
Molekylvægt: 237,29 g/mol
Opbevaringstemperatur: Køleskab
CAS-nummer: 2803455-97-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR