3-(2,4-Dioxo-3,4-dihydropyrimidin-1(2H)-yl)propanamide 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01446055-250EA 776 DKK
BLDPBD01446055-250 BLDPBD01446055-100
3-(2,4-Dioxo-3,4-dihydropyrimidin-1(2H)-yl)propanamide 97%
3-(2,4-Dioxo-3,4-dihydropyrimidin-1(2H)-yl)propanamide

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₉N₃O₃
Molekylvægt: 183,16 g/mol
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 89853-99-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR