(S)-4-Cyclohexyl-2-(6-cyclopropylpyridin-2-yl)-4,5-dihydrooxazole 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01445073-100EA 1350 DKK
BLDPBD01445073-100 BLDPBD01445073-250 BLDPBD01445073-1G
(S)-4-Cyclohexyl-2-(6-cyclopropylpyridin-2-yl)-4,5-dihydrooxazole 97%
(S)-4-Cyclohexyl-2-(6-cyclopropylpyridin-2-yl)-4,5-dihydrooxazole

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₇H₂₂N₂O
Molekylvægt: 270,36 g/mol
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 2828438-65-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR