2,2',2'',2'''-(Benzo[c][1,2,5]thiadiazole-4,7-diylbis([1,1':3',1''-terphenyl]-5',4,4''-triyl))tetraacetonitrile 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01444621-1GEA 4980 DKK
BLDPBD01444621-1G BLDPBD01444621-100 BLDPBD01444621-250
2,2',2'',2'''-(Benzo[c][1,2,5]thiadiazole-4,7-diylbis([1,1':3',1''-terphenyl]-5',4,4''-triyl))tetraacetonitrile 97%
2,2',2'',2'''-(Benzo[c][1,2,5]thiadiazole-4,7-diylbis([1,1':3',1''-terphenyl]-5',4,4''-triyl))tetraacetonitrile

Storage condition: Sealed in dry, Room Temperature

Formel: C₅₀H₃₂N₆S
Molekylvægt: 748,8943 g/mol
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 2803452-14-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR