4-Iodo-N-(4-(pentafluoro-l6-sulphanyl)phenyl)benzamide 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01444618-250EA 411 DKK
BLDPBD01444618-250 BLDPBD01444618-1G BLDPBD01444618-5G
4-Iodo-N-(4-(pentafluoro-l6-sulphanyl)phenyl)benzamide 95%
4-Iodo-N-(4-(pentafluoro-l6-sulphanyl)phenyl)benzamide

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₃H₉F₅INOS
Molekylvægt: 449,1782 g/mol
Opbevaringstemperatur: Køleskab
CAS-nummer: 2757085-55-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR