3-(4-(6-Methyl-1,2,4,5-tetrazin-3-yl)phenoxy)propan-1-amine hydrochloride 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01443569-50MEA 2780 DKK
BLDPBD01443569-50M BLDPBD01443569-100 BLDPBD01443569-25M
3-(4-(6-Methyl-1,2,4,5-tetrazin-3-yl)phenoxy)propan-1-amine hydrochloride 97%
3-(4-(6-Methyl-1,2,4,5-tetrazin-3-yl)phenoxy)propan-1-amine hydrochloride

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₂H₁₆ClN₅O
Molekylvægt: 281,74 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD31814446
CAS-nummer: 1802908-03-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR