(4R,5R)-1,3-Bis(2-isopropylphenyl)-4,5-diphenyl-4,5-dihydro-1H-imidazol-3-ium tetrafluoroborate 98%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD01442218-100EA
1680
DKK
BLDPBD01442218-100
BLDPBD01442218-250
(4R,5R)-1,3-Bis(2-isopropylphenyl)-4,5-diphenyl-4,5-dihydro-1H-imidazol-3-ium tetrafluoroborate 98%
(4R,5R)-1,3-Bis(2-isopropylphenyl)-4,5-diphenyl-4,5-dihydro-1H-imidazol-3-ium tetrafluoroborate
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Formel:
C₃₃H₃₅BF₄N₂ Molekylvægt: 546,44 g/mol Opbevaringstemperatur: Køleskab |
CAS-nummer:
372517-14-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...