2,7-Di(pyridin-4-yl)naphthalene 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01441992-1GEA 2250 DKK
BLDPBD01441992-1G BLDPBD01441992-250 BLDPBD01441992-100
2,7-Di(pyridin-4-yl)naphthalene 95%
2,7-Di(pyridin-4-yl)naphthalene

Storage condition: Sealed in dry, Room Temperature

Formel: C₂₀H₁₄N₂
Molekylvægt: 282,3386 g/mol
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 1219814-29-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR