[[2,2'-[(1R,2R)-1,2-Cyclohexanediylbis[(nitrilo-κN)methylidyne]]bis[4-bis(1,1-dimethylethyl)phenolato-κO]](2-)](4-methylbenzenesulphonato-κO)cobalt 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01441410-250EA 1060 DKK
BLDPBD01441410-250 BLDPBD01441410-1G BLDPBD01441410-100
[[2,2'-[(1R,2R)-1,2-Cyclohexanediylbis[(nitrilo-κN)methylidyne]]bis[4-bis(1,1-dimethylethyl)phenolato-κO]](2-)](4-methylbenzenesulphonato-κO)cobalt 97%
[[2,2'-[(1R,2R)-1,2-Cyclohexanediylbis[(nitrilo-κN)methylidyne]]bis[4-bis(1,1-dimethylethyl)phenolato-κO]](2-)](4-methylbenzenesulphonato-κO)cobalt

Storage condition: Inert atmosphere, 2-8°C

Opbevaringstemperatur: Køleskab
CAS-nummer: 2828438-42-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR