3-Isopropyl-6,7-dihydro-5H-cyclopenta[c]pyridin-4-amine 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01440168-1GEA 2020 DKK
BLDPBD01440168-1G BLDPBD01440168-100 BLDPBD01440168-250
3-Isopropyl-6,7-dihydro-5H-cyclopenta[c]pyridin-4-amine 97%
3-Isopropyl-6,7-dihydro-5H-cyclopenta[c]pyridin-4-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₁H₁₆N₂
Molekylvægt: 176,2581 g/mol
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 2409124-11-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR