2,3-Difluoro-1-methoxy-4-((4-(trans-4-propylcyclohexyl)phenyl)ethynyl)benzene 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01438967-1GEA 125 DKK
BLDPBD01438967-1G BLDPBD01438967-5G BLDPBD01438967-25G
2,3-Difluoro-1-methoxy-4-((4-(trans-4-propylcyclohexyl)phenyl)ethynyl)benzene 95%
2,3-Difluoro-1-methoxy-4-((4-(trans-4-propylcyclohexyl)phenyl)ethynyl)benzene

Storage condition: Sealed in dry, Room Temperature

Formel: C₂₄H₂₆F₂O
Molekylvægt: 368,45 g/mol
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 1383449-87-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR