2-(2-(2-((4-Methylbenzyl)oxy)ethoxy)ethoxy)ethanol 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01438937-100EA 334 DKK
BLDPBD01438937-100 BLDPBD01438937-1G BLDPBD01438937-250
2-(2-(2-((4-Methylbenzyl)oxy)ethoxy)ethoxy)ethanol 95%
2-(2-(2-((4-Methylbenzyl)oxy)ethoxy)ethoxy)ethanol

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₄H₂₂O₄
Molekylvægt: 254,32 g/mol
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 138471-93-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR