4,4'-(2,2-Bis(4-methoxyphenyl)ethene-1,1-diyl)dibenzaldehyde 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01437948-100EA 2080 DKK
BLDPBD01437948-100 BLDPBD01437948-50M BLDPBD01437948-1G BLDPBD01437948-250
4,4'-(2,2-Bis(4-methoxyphenyl)ethene-1,1-diyl)dibenzaldehyde 95%
4,4'-(2,2-Bis(4-methoxyphenyl)ethene-1,1-diyl)dibenzaldehyde

Storage condition: Inert atmosphere, Room Temperature

Formel: C₃₀H₂₄O₄
Molekylvægt: 448,5092 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD32708811
CAS-nummer: 2165348-28-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR