(S)-3-(4-Bromophenyl)piperidine oxalate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01437794-250EA 163 DKK
BLDPBD01437794-250 BLDPBD01437794-1G BLDPBD01437794-5G
(S)-3-(4-Bromophenyl)piperidine oxalate 95%
(S)-3-(4-Bromophenyl)piperidine oxalate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₃H₁₆BrNO₄
Molekylvægt: 330,1744 g/mol
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 2803376-59-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR