tert-Butyl 4-(3-chloropropyl)piperazine-1-carboxylate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01437246-100EA 311 DKK
BLDPBD01437246-100 BLDPBD01437246-5G BLDPBD01437246-1G BLDPBD01437246-250
tert-Butyl 4-(3-chloropropyl)piperazine-1-carboxylate 95%
tert-Butyl 4-(3-chloropropyl)piperazine-1-carboxylate

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₂H₂₃ClN₂O₂
Molekylvægt: 262,77 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD10697871
CAS-nummer: 165530-45-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR