Ethyl 3-hydroxy-4-nitro-1,5,6,7-tetrahydro-s-indacene-2-carboxylate 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01435543-250EA 745 DKK
BLDPBD01435543-250 BLDPBD01435543-100
Ethyl 3-hydroxy-4-nitro-1,5,6,7-tetrahydro-s-indacene-2-carboxylate 98%
Ethyl 3-hydroxy-4-nitro-1,5,6,7-tetrahydro-s-indacene-2-carboxylate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₅H₁₅NO₅
Molekylvægt: 289,28 g/mol
Opbevaringstemperatur: Køleskab
CAS-nummer: 2676865-16-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR