2-Methyl-1,2,3,5,6,7-hexahydro-s-indacen-4-amine 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01435243-100EA 876 DKK
BLDPBD01435243-100
2-Methyl-1,2,3,5,6,7-hexahydro-s-indacen-4-amine 97%
2-Methyl-1,2,3,5,6,7-hexahydro-s-indacen-4-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₃H₁₇N
Molekylvægt: 187,28 g/mol
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 2676867-60-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR