Ethyl 3-amino-7-methylbenzo[b]thiophene-2-carboxylate 97+%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01432922-250EA 2100 DKK
BLDPBD01432922-250 BLDPBD01432922-50M BLDPBD01432922-1G BLDPBD01432922-100
Ethyl 3-amino-7-methylbenzo[b]thiophene-2-carboxylate 97+%
Ethyl 3-amino-7-methylbenzo[b]thiophene-2-carboxylate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₁₂H₁₃NO₂S
Molekylvægt: 235,3021 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD29166946
CAS-nummer: 2092140-45-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR