5-Cyclopropyl-3-fluoropicolinaldehyde 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01431409-250EA 2890 DKK
BLDPBD01431409-250 BLDPBD01431409-1G BLDPBD01431409-100
5-Cyclopropyl-3-fluoropicolinaldehyde 95%
5-Cyclopropyl-3-fluoropicolinaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formel: C₉H₈FNO
Molekylvægt: 165,16 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD20228759
CAS-nummer: 1315363-79-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR