3-(4-Amino-2-nitrophenyl)propanoic acid hydrochloride 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01428408-100EA 1440 DKK
BLDPBD01428408-100 BLDPBD01428408-250 BLDPBD01428408-1G BLDPBD01428408-50M
3-(4-Amino-2-nitrophenyl)propanoic acid hydrochloride 97%
3-(4-Amino-2-nitrophenyl)propanoic acid hydrochloride

Storage condition: Sealed in dry, 2-8°C

Formel: C₉H₁₁ClN₂O₄
Molekylvægt: 246,64 g/mol
Opbevaringstemperatur: Køleskab
CAS-nummer: 2757730-30-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR