2-Cyano-6-fluorophenylboronic acid 98%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD01422364-5GEA
3280
DKK
BLDPBD01422364-5G
BLDPBD01422364-250
BLDPBD01422364-1G
2-Cyano-6-fluorophenylboronic acid 98%
2-Cyano-6-fluorophenylboronic acid
Storage condition: Inert atmosphere, 2-8°C
Formel:
C₇H₅BFNO₂ Molekylvægt: 164,9295 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD09033623 CAS-nummer: 656235-44-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...