2-Amino-5-bromobenzophenone 99+%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01400916-100EA 2140 DKK
BLDPBD01400916-100 BLDPBD01400916-25G BLDPBD01400916-5G
2-Amino-5-bromobenzophenone 99+%
2-Amino-5-bromobenzophenone

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₃H₁₀BrNO
Molekylvægt: 276,13 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00059962
CAS-nummer: 39859-36-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR