(4-Nitrophenyl)-β-D-galactopyranoside 96%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01387211-25GEA 931 DKK
BLDPBD01387211-25G BLDPBD01387211-1G BLDPBD01387211-5G
(4-Nitrophenyl)-β-D-galactopyranoside 96%
(4-Nitrophenyl)-β-D-galactopyranoside

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₂H₁₅NO₈
Molekylvægt: 301,25 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00063256
CAS-nummer: 3150-24-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR