3-(Morpholinosulphonyl)phenylboronic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01377508-1GEA 1160 DKK
BLDPBD01377508-1G BLDPBD01377508-5G BLDPBD01377508-250
3-(Morpholinosulphonyl)phenylboronic acid 98%
3-(Morpholinosulphonyl)phenylboronic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₀H₁₄BNO₅S
Molekylvægt: 271,09 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD07363748
CAS-nummer: 871329-60-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR