(2-Chlorobenzyl)triphenylphosphonium chloride 98%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01273116-5GEA 249 DKK
BLDPBD01273116-5G BLDPBD01273116-100 BLDPBD01273116-25G BLDPBD01273116-1G
(2-Chlorobenzyl)triphenylphosphonium chloride 98%
(2-Chlorobenzyl)triphenylphosphonium chloride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₂₅H₂₁Cl₂P
Molekylvægt: 423,3140 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00054629
CAS-nummer: 18583-55-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR