4-(Methylamino)phenol sulphate 99+%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: MetolBis(4-hydroxy-N-methylanilinium)sulfat

BLDPBD01124383-25GEA 140 DKK
BLDPBD01124383-25G BLDPBD01124383-5G
4-(Methylamino)phenol sulphate 99+%
Bis(4-hydroxy-N-methylanilinium)sulfat

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₄H₂₀N₂O₆S
Molekylvægt: 344,39 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00013140
CAS-nummer: 55-55-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR