Dibutyltin bis(2-ethylhexanoate) 97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD01090813-25GEA
62.1
DKK
BLDPBD01090813-25G
BLDPBD01090813-5G
Dibutyltin bis(2-ethylhexanoate) 97%
Dibutyltin bis(2-ethylhexanoate)
Storage condition: Sealed in dry, Room Temperature
Formel:
C₂₄H₄₈O₄Sn Molekylvægt: 519,34 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00015261 CAS-nummer: 2781-10-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...