3-Chloro-p-toluidine hydrochloride 95%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD00852485-100EA
2270
DKK
BLDPBD00852485-100
BLDPBD00852485-1G
BLDPBD00852485-25G
BLDPBD00852485-5G
3-Chloro-p-toluidine hydrochloride 95%
3-Chloro-p-toluidine hydrochloride
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₇H₉Cl₂N Molekylvægt: 178,06 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00242925 CAS-nummer: 7745-89-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...