3'-Fluoro-4'-nitroacetanilide 98%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD00825849-5GEA 1480 DKK
BLDPBD00825849-5G BLDPBD00825849-1G BLDPBD00825849-250
3'-Fluoro-4'-nitroacetanilide 98%
3'-Fluoro-4'-nitroacetanilide

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₇FN₂O₃
Molekylvægt: 198,1512 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01463345
CAS-nummer: 345-30-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR