7-Bromo-2,3-dihydro-2,2-dimethyl-1H-carbazol-4(9H)-one 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD00803179-100EA 3150 DKK
BLDPBD00803179-100 BLDPBD00803179-250
7-Bromo-2,3-dihydro-2,2-dimethyl-1H-carbazol-4(9H)-one 95%
7-Bromo-2,3-dihydro-2,2-dimethyl-1H-carbazol-4(9H)-one

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₄H₁₄BrNO
Molekylvægt: 292,171 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD29765459
CAS-nummer: 1301219-07-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR